Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFMBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016329.3 | NP_057413.2 | 6 | 967 | ||
XM_005265221.3 | XP_005265278.1 | 6 | 1083 | |||
XM_006713203.3 | XP_006713266.1 | 7 | 1205 | |||
XM_006713204.3 | XP_006713267.1 | 6 | 905 | |||
XM_011533824.2 | XP_011532126.1 | 7 | 1081 | |||
XM_011533825.2 | XP_011532127.1 | 7 | 928 | |||
GenBank mRNA | AB189472.1 | - | 5 | 591 | ||
AF168132.1 | - | 6 | 904 | |||
AK313965.1 | - | 6 | 962 | |||
AL080140.1 | - | 3 | 245 | |||
BC014614.2 | - | 6 | 851 | 3029598 |