Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 9 | 1757 | ||
NM_013289.2 | NP_037421.2 | 9 | 1757 | |||
GenBank mRNA | BC028206.1 | - | 9 | 1721 | 5215674 | |
L41269.1 | - | 9 | 1713 | |||
L76664.1 | - | 8 | 1420 | |||
U30273.1 | - | 9 | 1696 | |||
U30274.1 | - | 9 | 1757 | |||
U31416.1 | - | 9 | 1719 | |||
U33328.1 | - | 8 | 1668 |