Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205313.1 | NP_001192242.1 | 22 | 2221 | ||
NM_001205314.1 | NP_001192243.1 | 22 | 2239 | |||
NM_009283.4 | NP_033309.3 | 22 | 2254 | |||
GenBank mRNA | AK018544.1 | - | 22 | 2205 | ||
AK039458.1 | - | 22 | 2255 | |||
AK046517.1 | - | 22 | 2221 | |||
AK082706.1 | - | 22 | 2236 | |||
AK084855.1 | - | 22 | 2188 | |||
AK157263.1 | - | 22 | 2103 | |||
AK159887.1 | - | 22 | 2238 | |||
BC042551.1 | - | 5 | 386 | |||
U06924.1 | - | 19 | 1908 |