Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POU5F1P4 SIRNA Assays ›
Gene Symbol:
POU5F1P4
POU5F1P3
Gene Name:
POU class 5 homeobox 1 pseudogene 4
POU class 5 homeobox 1 pseudogene 3
Gene Aliases:
Oct4-pg4, POU5FLC1
OCT4-pg3, OTF3L, POU5F1L, POU5FLC12
Chromosome Location:
Chr.1: 155433180 - 155434262 on Build GRCh38
Chromosome Location:
Chr.12: 8133769 - 8134852 on Build GRCh38
UniGene ID:
Hs.632482
Hs.646545
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
241322
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POU5F1P4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_034180.1
1
333
GenBank mRNA
AF268613.1
-
1
333
DQ851566.1
-
1
333
GU480814.1
-
1
334
GU480831.1
-
1
335
GU480833.1
-
1
334
GU480834.1
-
1
334
GU480836.1
-
1
334
GU480839.1
-
1
334
GU480848.1
-
1
334
GU480852.1
-
1
334
GU480853.1
-
1
335
GU480854.1
-
1
334
GU480855.1
-
1
332
GU480856.1
-
1
334
GU480857.1
-
1
334
GU480858.1
-
1
334
GU480859.1
-
1
334
GU480860.1
-
1
333
GU480861.1
-
1
335
GU480862.1
-
1
334
GU480863.1
-
1
334
GU480865.1
-
1
335
GU480866.1
-
1
334
GU480867.1
-
1
334
GU480869.1
-
1
334
GU480870.1
-
1
334
GU480871.1
-
1
334
GU480872.1
-
1
334
GU480873.1
-
1
334
GU480874.1
-
1
334
GU480875.1
-
1
334
GU480879.1
-
1
334
GU480881.1
-
1
334
GU480882.1
-
1
334
GU480890.1
-
1
333
GU480894.1
-
1
334
GU480895.1
-
1
334
GU480898.1
-
1
333
GU480900.1
-
1
333
GU480904.1
-
1
334
GU480914.1
-
1
333
HQ122676.1
-
1
334
HQ122677.1
-
1
334
HQ122678.1
-
1
332
HQ122679.1
-
1
334
HQ122680.1
-
1
334
HQ122681.1
-
1
334
HQ122682.1
-
1
334
HQ122683.1
-
1
335
HQ122684.1
-
1
334
HQ122685.1
-
1
332
HQ122686.1
-
1
334
HQ122687.1
-
1
334
HQ122688.1
-
1
333
HQ122689.1
-
1
332
HQ122690.1
-
1
334
HQ122691.1
-
1
334
HQ122695.1
-
1
333
HQ122696.1
-
1
333
HQ122697.1
-
1
333
HQ122698.1
-
1
334
HQ122699.1
-
1
334
HQ122700.1
-
1
334
HQ122701.1
-
1
334
HQ122702.1
-
1
334
HQ122703.1
-
1
334
HQ122704.1
-
1
334
HQ122705.1
-
1
334
HQ122706.1
-
1
334
HQ122707.1
-
1
333
HQ122708.1
-
1
334
HQ122709.1
-
1
334
HQ122731.1
-
1
334
HQ122732.1
-
1
334
HQ122733.1
-
1
334
HQ122734.1
-
1
334
HQ122735.1
-
1
334
HQ122736.1
-
1
334
HQ122737.1
-
1
334
HQ122738.1
-
1
334
HQ122739.1
-
1
334
HQ122740.1
-
1
334
HQ122741.1
-
1
334
HQ122742.1
-
1
334
HQ122743.1
-
1
334
HQ122744.1
-
1
334
HQ122745.1
-
1
334
HQ122746.1
-
1
334
HQ122747.1
-
1
334
HQ122748.1
-
1
334
HQ122756.1
-
1
334
HQ122757.1
-
1
334
HQ122758.1
-
1
334
HQ122759.1
-
1
334
HQ122760.1
-
1
334
HQ122761.1
-
1
334
HQ122762.1
-
1
334
HQ122763.1
-
1
334
HQ122764.1
-
1
334
HQ122765.1
-
1
334
HQ122766.1
-
1
334
HQ122767.1
-
1
334
HQ122768.1
-
1
334
HQ122769.1
-
1
334
HQ122770.1
-
1
334
HQ122771.1
-
1
334
HQ122772.1
-
1
334
HQ122773.1
-
1
334
HQ122774.1
-
1
334
HQ122775.1
-
1
334
HQ122776.1
-
1
334
HQ122777.1
-
1
334
HQ122778.1
-
1
334
HQ122779.1
-
1
334
HQ122780.1
-
1
334
HQ122798.1
-
1
334
HQ122799.1
-
1
333
HQ122800.1
-
1
334
HQ122801.1
-
1
334
HQ122802.1
-
1
332
HQ122804.1
-
1
334
HQ122805.1
-
1
334
HQ122806.1
-
1
334
HQ122807.1
-
1
334
HQ122808.1
-
1
334
HQ122809.1
-
1
332
HQ122810.1
-
1
334
HQ122811.1
-
1
334
HQ122812.1
-
1
334
HQ122813.1
-
1
334
HQ122814.1
-
1
332
HQ122815.1
-
1
334
HQ122832.1
-
1
334
HQ122833.1
-
1
334
HQ122834.1
-
1
334
HQ122835.1
-
1
332
HQ122836.1
-
1
334
HQ122837.1
-
1
334
HQ122838.1
-
1
332
HQ122839.1
-
1
334
HQ122840.1
-
1
334
HQ122858.1
-
1
334
HQ122859.1
-
1
334
HQ122861.1
-
1
334
HQ122862.1
-
1
334
HQ122888.1
-
1
334
HQ122889.1
-
1
334
HQ122890.1
-
1
334
HQ122904.1
-
1
334
HQ122905.1
-
1
334
HQ122906.1
-
1
334
HQ122907.1
-
1
334
HQ122908.1
-
1
334
HQ122909.1
-
1
335
HQ122910.1
-
1
334
HQ122911.1
-
1
334
HQ122912.1
-
1
334
HQ122913.1
-
1
334
HQ122914.1
-
1
334
Gene Symbol: POU5F1P3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
GU480915.1
-
1
333
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0