Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164814.1 | NP_001158286.1 | 12 | 2976 | ||
NM_001164815.1 | NP_001158287.1 | 11 | 2895 | |||
NM_001164816.1 | NP_001158288.1 | 5 | 534 | |||
NM_001164817.1 | NP_001158289.1 | 6 | 596 | |||
NM_014977.3 | NP_055792.1 | 12 | 3015 | |||
XM_005267415.3 | XP_005267472.1 | 12 | 2816 | |||
XM_005267416.3 | XP_005267473.1 | 12 | 2783 | |||
XM_005267418.1 | XP_005267475.1 | 5 | 831 | |||
GenBank mRNA | AB014570.2 | - | 12 | 2524 | ||
AF124726.1 | - | 12 | 3014 | |||
AF124727.1 | - | 6 | 550 | |||
AF124728.1 | - | 5 | 525 | |||
AK299034.1 | - | 11 | 2406 | |||
AL050382.1 | - | 5 | 1620 | |||
AL833228.1 | - | 7 | 865 | |||
BC140805.1 | - | 12 | 2745 | 9021673 | ||
BX247975.1 | - | 5 | 804 | |||
CX786860.1 | - | 6 | 528 |