Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DOCK9 SIRNA Assays ›
Gene Symbol:
DOCK9
Gene Name:
dedicator of cytokinesis 9
Gene Aliases:
ZIZ1, ZIZIMIN1
Chromosome Location:
Chr.13: 98793487 - 99086406 on Build GRCh38
UniGene ID:
Hs.596105
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241602
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DOCK9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130048.1
NP_001123520.1
44
5107
NM_001318849.1
NP_001305778.1
44
4990
NM_015296.2
NP_056111.1
44
4990
XM_005254034.3
XP_005254091.1
44
5048
XM_005254035.3
XP_005254092.1
44
4955
XM_006719922.1
XP_006719985.1
44
5209
XM_006719924.2
XP_006719987.1
44
5048
XM_006719925.1
XP_006719988.1
44
5176
XM_006719931.3
XP_006719994.1
44
4935
XM_006719932.1
XP_006719995.1
44
5107
XM_006719936.1
XP_006719999.1
44
5140
XM_006719937.3
XP_006720000.1
44
4935
XM_017020503.1
XP_016875992.1
44
5049
XM_017020504.1
XP_016875993.1
44
5049
XM_017020505.1
XP_016875994.1
44
5049
XM_017020506.1
XP_016875995.1
44
4956
XM_017020507.1
XP_016875996.1
44
5049
XM_017020508.1
XP_016875997.1
44
4956
XM_017020509.1
XP_016875998.1
44
5176
XM_017020510.1
XP_016875999.1
44
4956
XM_017020511.1
XP_016876000.1
44
4980
XM_017020512.1
XP_016876001.1
44
5176
XM_017020513.1
XP_016876002.1
44
4956
XM_017020514.1
XP_016876003.1
44
5176
XM_017020515.1
XP_016876004.1
44
5083
XM_017020516.1
XP_016876005.1
44
5176
XM_017020517.1
XP_016876006.1
44
5055
XM_017020518.1
XP_016876007.1
44
5079
XM_017020519.1
XP_016876008.1
44
5083
XM_017020520.1
XP_016876009.1
44
5083
XM_017020521.1
XP_016876010.1
44
5140
GenBank mRNA
AB028981.2
-
44
5013
AF527605.1
-
44
4990
AK090793.1
-
8
2046
AK127215.1
-
7
2079
AK127329.1
-
45
5116
AK294573.1
-
10
1032
AK294705.1
-
17
1830
AK294770.1
-
7
1169
AK300298.1
-
9
1061
AK300331.1
-
10
1147
BC043506.1
-
3
286
BC053620.1
-
4
455
BC136274.1
-
44
4970
9020261
BC171753.1
-
44
4970
9053254
BX648707.1
-
7
2872
CR749658.1
-
7
1973
CR936820.1
-
43
9419
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
blood coagulation
biological_process
positive regulation of GTPase activity
cell-cell adhesion
guanyl-nucleotide exchange factor activity
protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-sjrww:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0