Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IP6K2 SIRNA Assays ›
Gene Symbol:
IP6K2
Gene Name:
inositol hexakisphosphate kinase 2
Gene Aliases:
IHPK2, PIUS
Chromosome Location:
Chr.3: 48688003 - 48717278 on Build GRCh38
UniGene ID:
Hs.595983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24192
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IP6K2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005909.2
NP_001005909.1
2
397
NM_001005910.2
NP_001005910.1
2
400
NM_001005911.2
NP_001005911.1
2
397
NM_001146178.2
NP_001139650.1
2
400
NM_001146179.2
NP_001139651.1
2
397
NM_001190316.1
NP_001177245.1
3
613
NM_001190317.1
NP_001177246.1
3
536
NM_016291.3
NP_057375.2
2
400
NR_027437.2
2
400
NR_027438.2
2
397
XM_006713199.3
XP_006713262.1
3
504
XM_006713200.1
XP_006713263.1
3
474
XM_006713201.1
XP_006713264.1
3
551
XM_006713202.1
XP_006713265.1
3
553
XM_011533816.1
XP_011532118.1
3
534
XM_011533817.1
XP_011532119.1
2
337
XM_011533818.1
XP_011532120.1
2
340
XM_011533822.1
XP_011532124.1
2
337
XM_011533823.1
XP_011532125.1
2
340
XM_017006583.1
XP_016862072.1
2
396
XM_017006584.1
XP_016862073.1
2
371
XM_017006585.1
XP_016862074.1
3
486
XM_017006586.1
XP_016862075.1
3
527
XM_017006587.1
XP_016862076.1
3
527
XM_017006588.1
XP_016862077.1
3
553
XM_017006589.1
XP_016862078.1
3
502
XM_017006590.1
XP_016862079.1
3
1108
XM_017006591.1
XP_016862080.1
3
473
XM_017006592.1
XP_016862081.1
3
502
XM_017006593.1
XP_016862082.1
3
502
GenBank mRNA
AF177145.1
-
1
294
AK024045.1
-
2
337
AK097016.1
-
8
1255
AK290526.1
-
3
551
AK291501.1
-
3
528
AK304714.1
-
3
474
AK315204.1
-
2
338
AL137514.1
-
2
338
AY007091.1
-
2
323
BC001864.1
-
2
322
BC004469.2
-
2
327
BC019694.1
-
2
309
4941646
BC065533.1
-
2
326
6162048
BP262706.1
-
2
337
BX537544.1
-
3
552
CA488567.1
-
2
334
CD248129.1
-
2
363
CF139656.1
-
2
331
CN362780.1
-
2
312
DA083858.1
-
2
333
DB039955.1
-
2
397
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
phosphate ion transport
negative regulation of cell growth
positive regulation of apoptotic process
inositol phosphate metabolic process
phosphatidylinositol phosphorylation
type I interferon signaling pathway
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
protein binding
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0