Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPH7 SIRNA Assays ›
Gene Symbol:
DPH7
Gene Name:
diphthamide biosynthesis 7
Gene Aliases:
C9orf112, RRT2, WDR85
Chromosome Location:
Chr.9: 137554899 - 137578935 on Build GRCh38
UniGene ID:
Hs.292570
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241940
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPH7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138778.3
NP_620133.1
9
1289
XM_005266123.2
XP_005266180.1
10
1747
XM_005266126.3
XP_005266183.1
8
1520
XM_005266128.2
XP_005266185.1
10
1373
XM_005266129.2
XP_005266186.1
10
1376
XM_005266130.2
XP_005266187.1
8
1144
XM_011519188.1
XP_011517490.1
8
1212
XM_011519189.1
XP_011517491.1
9
1390
XM_011519191.2
XP_011517493.1
9
2445
XM_011519193.2
XP_011517495.1
9
2494
XM_011519199.2
XP_011517501.1
9
1235
XM_011519200.1
XP_011517502.1
9
1238
XM_017015289.1
XP_016870778.1
8
2379
XM_017015290.1
XP_016870779.1
8
2428
XM_017015291.1
XP_016870780.1
10
1582
XM_017015292.1
XP_016870781.1
9
1652
XM_017015293.1
XP_016870782.1
9
1609
XM_017015294.1
XP_016870783.1
11
1562
XM_017015295.1
XP_016870784.1
9
1649
XM_017015296.1
XP_016870785.1
9
1488
XM_017015297.1
XP_016870786.1
11
1559
XM_017015298.1
XP_016870787.1
10
1842
XM_017015299.1
XP_016870788.1
10
1099
XM_017015300.1
XP_016870789.1
10
1839
XM_017015301.1
XP_016870790.1
8
1512
XM_017015302.1
XP_016870791.1
8
2566
XM_017015303.1
XP_016870792.1
9
1747
XM_017015304.1
XP_016870793.1
8
1212
XM_017015305.1
XP_016870794.1
8
2543
XM_017015306.1
XP_016870795.1
9
1681
XM_017015307.1
XP_016870796.1
9
1678
XM_017015308.1
XP_016870797.1
7
2405
XM_017015309.1
XP_016870798.1
9
1405
GenBank mRNA
AJ420445.1
-
1
648
AK075115.1
-
9
1290
AK094389.1
-
1
1290
BC003123.1
-
9
1228
3502107
BC017335.2
-
9
1248
4642600
BC040173.1
-
2
751
4791980
Back To Top
More Information
Panther Classification:
Molecular Function -
methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
diphthine methylesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0