Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016245.4 | NP_057329.3 | 7 | 1572 | ||
GenBank mRNA | AF126780.1 | - | 7 | 1527 | ||
AF273056.1 | - | 7 | 1425 | |||
AK075348.1 | - | 7 | 1456 | |||
AY358553.1 | - | 7 | 1439 | |||
BC036001.1 | - | 7 | 1463 | 4715042 | ||
CB163647.1 | - | 1 | 247 |