Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243042.1 | NP_001229971.1 | 8 | 1460 | ||
NM_002117.5 | NP_002108.4 | 8 | 1460 | |||
GenBank mRNA | AK024836.1 | - | 6 | 1325 | ||
AK130592.1 | - | 9 | 1326 | |||
AK304527.1 | - | 8 | 1272 | |||
BC007814.2 | - | 8 | 1393 | |||
BC033293.1 | - | 8 | 1402 | 4780246 | ||
M11886.1 | - | 7 | 1323 | |||
M26429.1 | - | 8 | 1393 | |||
M28207.1 | - | 7 | 1165 | |||
X58536.1 | - | 9 | 1409 |