Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012412.4 | NP_036544.1 | 4 | 391 | ||
NM_138635.3 | NP_619541.1 | 4 | 391 | |||
NM_201436.2 | NP_958844.1 | 3 | 313 | |||
NM_201517.2 | NP_958925.1 | 3 | 277 | |||
GenBank mRNA | AB209001.1 | - | 4 | 302 | ||
AF081192.1 | - | 4 | 256 | |||
AK023973.1 | - | 4 | 342 | |||
AK129989.1 | - | 4 | 311 | |||
AL520785.1 | - | 1 | 39 | |||
AL520811.3 | - | 4 | 292 | |||
BC000098.2 | - | 4 | 318 | 3505930 | ||
BC004274.1 | - | 3 | 243 | 3615320 | ||
BC014885.2 | - | 4 | 287 | 3907411 | ||
BC070169.1 | - | 4 | 318 | 6458566 | ||
BG754026.1 | - | 4 | 366 | |||
BG773256.1 | - | 4 | 394 | |||
BI257891.1 | - | 4 | 349 | |||
BM452844.1 | - | 4 | 272 | |||
BM453663.1 | - | 4 | 261 |