Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028496.1 | 3 | 513 | |||
GenBank mRNA | BC024224.1 | - | 3 | 517 | 3998034 |