Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289080.1 | NP_001276009.1 | 6 | 1018 | ||
NM_001289081.1 | NP_001276010.1 | 5 | 881 | |||
NM_014461.3 | NP_055276.1 | 6 | 1027 | |||
XM_005265058.3 | XP_005265115.1 | 7 | 1150 | |||
XM_011533590.2 | XP_011531892.1 | 6 | 674 | |||
XM_011533591.2 | XP_011531893.1 | 5 | 989 | |||
XM_017006171.1 | XP_016861660.1 | 11 | 1577 | |||
XM_017006172.1 | XP_016861661.1 | 6 | 1152 | |||
XM_017006173.1 | XP_016861662.1 | 6 | 766 | |||
XM_017006174.1 | XP_016861663.1 | 5 | 641 | |||
XM_017006175.1 | XP_016861664.1 | 4 | 418 | |||
XM_017006177.1 | XP_016861666.1 | 6 | 766 | |||
GenBank mRNA | AB003592.1 | - | 6 | 740 | ||
AK294786.1 | - | 5 | 881 | |||
AK307814.1 | - | 7 | 767 | |||
AK314220.1 | - | 6 | 741 | |||
BC113118.1 | - | 6 | 653 | 40036214 |