Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZXDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007156.4 | NP_009087.1 | 1 | 2483 | ||
GenBank mRNA | AK122815.1 | - | 1 | 2482 | ||
AK292686.1 | - | 1 | 2473 | |||
BC059356.1 | - | 1 | 2281 | 30347480 | ||
L14787.1 | - | 1 | 1365 |