Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016436.4 | NP_057520.2 | 3 | 236 | ||
XM_011528843.1 | XP_011527145.1 | 3 | 225 | |||
XM_017027864.1 | XP_016883353.1 | 4 | 318 | |||
XM_017027865.1 | XP_016883354.1 | 4 | 519 | |||
XM_017027866.1 | XP_016883355.1 | 3 | 439 | |||
XM_017027867.1 | XP_016883356.1 | 3 | 274 | |||
XM_017027868.1 | XP_016883357.1 | 3 | 225 | |||
XM_017027869.1 | XP_016883358.1 | 4 | 313 | |||
XM_017027870.1 | XP_016883359.1 | 4 | 319 | |||
XM_017027871.1 | XP_016883360.1 | 4 | 319 | |||
GenBank mRNA | AF220416.1 | - | 3 | 200 | ||
AF258787.1 | - | 3 | 222 | |||
AF348207.1 | - | 2 | 161 | |||
AK054597.1 | - | 4 | 493 | |||
AK314503.1 | - | 3 | 200 | |||
AL558423.3 | - | 2 | 159 | |||
AY027523.1 | - | 2 | 161 | |||
AY134747.1 | - | 3 | 213 | |||
BC006415.1 | - | 3 | 176 | |||
BC016948.1 | - | 3 | 189 | |||
BC048210.1 | - | 4 | 269 | |||
BC080598.1 | - | 3 | 200 | |||
BC093405.1 | - | 3 | 236 | |||
BC150178.1 | - | 3 | 202 | 40148074 | ||
BG778940.1 | - | 4 | 306 | |||
BM906582.1 | - | 3 | 225 | |||
BX643875.1 | - | 3 | 194 | |||
CD690482.1 | - | 3 | 238 | |||
L09749.1 | - | 2 | 115 |