Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288792.1 | NP_001275721.1 | 3 | 394 | ||
NM_198481.3 | NP_940883.2 | 3 | 394 | |||
NR_110142.1 | 3 | 394 | ||||
XM_011526845.1 | XP_011525147.1 | 3 | 407 | |||
XM_011526846.1 | XP_011525148.1 | 3 | 407 | |||
XM_011526847.1 | XP_011525149.1 | 3 | 407 | |||
XM_011526848.1 | XP_011525150.1 | 3 | 407 | |||
XM_011526849.1 | XP_011525151.1 | 3 | 407 | |||
XM_017026666.1 | XP_016882155.1 | 3 | 407 | |||
GenBank mRNA | AY358542.1 | - | 3 | 403 | ||
BC100943.2 | - | 3 | 371 | 40004095 | ||
DQ479397.1 | - | 3 | 235 | |||
FJ584316.1 | - | 3 | 229 | |||
FJ584317.1 | - | 3 | 229 | |||
FJ584318.1 | - | 3 | 229 | |||
FJ882051.1 | - | 3 | 229 | |||
FN398145.1 | - | 3 | 222 |