Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005018.2 | NP_005009.2 | 4 | 672 | ||
XM_017004293.1 | XP_016859782.1 | 4 | 674 | |||
GenBank mRNA | AK313848.1 | - | 4 | 674 | ||
AY206416.1 | - | 2 | 278 | |||
AY238517.1 | - | 4 | 604 | |||
AY322430.1 | - | 394 | ||||
BC074740.2 | - | 4 | 650 | 30915198 | ||
KJ865859.1 | - | 4 | 604 | |||
KJ865860.1 | - | 4 | 604 | |||
KJ865861.1 | - | 4 | 604 | |||
U64863.1 | - | 4 | 672 |