Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286272.1 | NP_001273201.1 | 3 | 333 | ||
NM_001286273.1 | NP_001273202.1 | 2 | 259 | |||
NM_003295.3 | NP_003286.1 | 3 | 333 | |||
GenBank mRNA | AK130847.1 | - | 3 | 232 | ||
AK296587.1 | - | 3 | 232 | |||
AK296952.1 | - | 2 | 759 | |||
AK312951.1 | - | 3 | 232 | |||
AY117678.1 | - | 3 | 233 | |||
AY334563.1 | - | 3 | 118 | |||
BC003352.2 | - | 3 | 216 | 2899964 | ||
BC012431.1 | - | 3 | 192 | 3880559 | ||
BC022436.1 | - | 2 | 187 | 4281364 | ||
BC040008.1 | - | 2 | 129 | |||
BC052333.1 | - | 3 | 222 | 6058799 | ||
BP215975.1 | - | 3 | 333 | |||
CR457036.1 | - | 3 | 118 | |||
X16064.1 | - | 3 | 212 | |||
X64899.1 | - | 3 | 204 |
Gene Symbol: FLJ44635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207422.2 | NP_997305.2 | 2 | 481 | ||
GenBank mRNA | AK126598.1 | - | 2 | 481 | ||
AY572224.1 | - | 2 | 481 | |||
BC137552.1 | - | 2 | 237 | 9021552 | ||
BC137564.1 | - | 2 | 237 | 9021564 |