Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079520.1 | NP_001072988.1 | 1 | 472 | ||
NM_016651.5 | NP_057735.2 | 1 | 472 | |||
GenBank mRNA | AI803159.1 | - | 1 | 367 | ||
CN304224.1 | - | 1 | 472 | |||
DR001289.1 | - | 1 | 455 |