Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001140.3 | NP_001131.3 | 6 | 682 | ||
GenBank mRNA | AK290309.1 | - | 6 | 695 | ||
AK296821.1 | - | 7 | 767 | |||
AK296904.1 | - | 6 | 578 | |||
AK312557.1 | - | 6 | 695 | |||
AK316126.1 | - | 6 | 838 | |||
BC029032.1 | - | 6 | 682 | 5179700 | ||
M23892.1 | - | 6 | 671 |
Gene Symbol: ALOX15P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045985.1 | 3 | 210 | |||
GenBank mRNA | DY655533.1 | - | 3 | 210 |