Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040437.2 | NP_001035527.1 | 4 | 453 | ||
NM_001040438.2 | NP_001035528.1 | 3 | 359 | |||
NM_001287482.1 | NP_001274411.1 | 4 | 464 | |||
NM_001287483.1 | NP_001274412.1 | 2 | 318 | |||
NM_001287484.1 | NP_001274413.1 | 3 | 363 | |||
NM_001287485.1 | NP_001274414.1 | 4 | 310 | |||
NM_001287486.1 | NP_001274415.1 | 4 | 307 | |||
NM_001287487.1 | NP_001274416.1 | 3 | 216 | |||
NM_001287488.1 | NP_001274417.1 | 3 | 104 | |||
GenBank mRNA | AJ249731.1 | - | 3 | 368 | ||
AJ249732.1 | - | 2 | 327 | |||
AK311989.1 | - | 2 | 317 | |||
AL521625.3 | - | 4 | 439 | |||
AL556045.3 | - | 4 | 419 | |||
BC000706.1 | - | 3 | 340 | 3349773 | ||
BC017660.1 | - | 4 | 257 | 4049404 | ||
BC066330.1 | - | 3 | 358 | |||
BG109213.1 | - | 4 | 381 | |||
BG116666.1 | - | 4 | 274 | |||
BJ996767.1 | - | 2 | 318 | |||
BM690817.1 | - | 3 | 178 | |||
BQ920527.1 | - | 4 | 437 | |||
CD247791.1 | - | 4 | 326 | |||
CD249104.1 | - | 3 | 374 | |||
HY060373.1 | - | 3 | 102 | |||
W52524.1 | - | 3 | 348 |