Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 6 | 1148 | ||
NM_201400.3 | NP_958802.1 | 8 | 1331 | |||
NM_201598.3 | NP_963892.1 | 7 | 1229 | |||
XM_005255157.4 | XP_005255214.1 | 7 | 1223 | |||
XM_005255158.4 | XP_005255215.1 | 9 | 1377 | |||
XM_011522404.2 | XP_011520706.1 | 8 | 1296 | |||
GenBank mRNA | BI491350.1 | - | 1 | 247 |
Gene Symbol: ALG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CD366777.1 | - | 1 | 36 |
Gene Symbol: FAM86C2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024249.1 | 4 | 690 | |||
GenBank mRNA | AK055771.1 | - | 2 | 1343 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN481473.1 | - | 3 | 516 |
Gene Symbol: FAM86FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024254.1 | 5 | 942 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 5 | 897 | |||
GenBank mRNA | AK124448.1 | - | 3 | 666 |
Gene Symbol: FAM86JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024250.1 | 5 | 771 | |||
NR_024251.1 | 5 | 869 | ||||
GenBank mRNA | BC032772.1 | - | 5 | 757 | ||
BC041080.1 | - | 5 | 840 |