Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3PXD2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038940.1 | 3 | 880 | |||
GenBank mRNA | AK056784.1 | - | 3 | 662 |
Gene Symbol: CCDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032040.4 | NP_114429.2 | 1 | 2683 | ||
GenBank mRNA | AL136609.1 | - | 1 | 2609 | ||
BC025243.1 | - | 1 | 2054 | 4553965 | ||
BX096201.1 | - | 1 | 46 | |||
JN703457.1 | - | 1 | 2605 |
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128234.1 | - | 1 | 453 |
Gene Symbol: EEF2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013302.3 | NP_037434.1 | 18 | 6548 | ||
XM_017023193.1 | XP_016878682.1 | 18 | 6259 | |||
XM_017023194.1 | XP_016878683.1 | 17 | 6492 |
Gene Symbol: ERLIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127017.1 | - | 1 | 290 |
Gene Symbol: FAM210A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098801.1 | NP_001092271.1 | 5 | 3958 | ||
NM_152352.3 | NP_689565.2 | 4 | 3818 | |||
GenBank mRNA | U78519.1 | - | 1 | 238 |
Gene Symbol: LOC100129550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024618.1 | 1 | 318 | |||
GenBank mRNA | BX471201.1 | - | 1 | 158 | ||
BX490907.1 | - | 1 | 324 |
Gene Symbol: LOC100507236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129682.1 | - | 1 | 402 |
Gene Symbol: LZTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021020.3 | NP_066300.1 | 3 | 3346 | ||
XM_005273394.3 | XP_005273451.1 | 4 | 3611 | |||
XM_011544383.2 | XP_011542685.1 | 4 | 5549 | |||
XM_011544384.2 | XP_011542686.1 | 4 | 3499 | |||
XM_011544385.2 | XP_011542687.1 | 4 | 3473 | |||
XM_011544386.2 | XP_011542688.1 | 4 | 3698 | |||
GenBank mRNA | AF123659.1 | - | 3 | 3346 |
Gene Symbol: NSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA811439.1 | - | 1 | 263 |
Gene Symbol: PLS3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110383.1 | 3 | 272 | |||
GenBank mRNA | DB018483.1 | - | 3 | 248 |
Gene Symbol: POLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278255.1 | NP_001265184.1 | 5 | 1805 | ||
NM_017443.4 | NP_059139.3 | 5 | 1835 | |||
NR_027261.1 | 4 | 1969 | ||||
GenBank mRNA | AF070640.1 | - | 1 | 1156 | ||
AK074762.1 | - | 5 | 1758 | |||
AK074782.1 | - | 4 | 1668 | |||
AK092840.1 | - | 3 | 1963 | |||
BC003166.1 | - | 4 | 1656 | 2822216 | ||
BC004170.2 | - | 4 | 1656 | 2822216 |
Gene Symbol: RMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024945.2 | NP_079221.2 | 3 | 2528 | ||
XM_005252211.2 | XP_005252268.1 | 3 | 2397 | |||
XM_005252213.2 | XP_005252270.1 | 4 | 2473 | |||
XM_011519034.2 | XP_011517336.1 | 4 | 2586 | |||
XM_017015140.1 | XP_016870629.1 | 3 | 2338 | |||
GenBank mRNA | AK022950.1 | - | 3 | 2450 | ||
BC039999.2 | - | 3 | 2353 | 5554889 |
Gene Symbol: SERAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032861.3 | NP_116250.3 | 17 | 3129 | ||
XM_006715586.2 | XP_006715649.1 | 16 | 3058 | |||
XM_011536196.2 | XP_011534498.1 | 16 | 3152 | |||
XM_011536198.2 | XP_011534500.1 | 15 | 3054 | |||
GenBank mRNA | AK023656.1 | - | 1 | 1165 | ||
BC001705.1 | - | 17 | 3086 | 3350731 | ||
BX648104.1 | - | 15 | 4506 |
Gene Symbol: TAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029399.1 | XP_016884888.1 | 10 | 2583 | ||
XM_017029400.1 | XP_016884889.1 | 10 | 2438 | |||
GenBank mRNA | AL832071.1 | - | 9 | 5823 |
Gene Symbol: VCPKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021640.1 | XP_016877129.1 | 6 | 5910 | ||
XM_017021641.1 | XP_016877130.1 | 6 | 5907 |