Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023029.1 | - | 5 | 1441 |
Gene Symbol: ALPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008644.1 | XP_016864133.1 | 1 | 1062 |
Gene Symbol: ASCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124364.1 | - | 1 | 65 |
Gene Symbol: ATP13A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032279.3 | NP_115655.2 | 30 | 6755 |
Gene Symbol: C12orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056213.1 | - | 4 | 2209 |
Gene Symbol: CEP57L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266935.4 | XP_005266992.3 | 11 | 5025 | ||
XM_005266936.4 | XP_005266993.3 | 11 | 5025 | |||
XM_005266937.4 | XP_005266994.3 | 11 | 4977 | |||
XM_005266938.4 | XP_005266995.3 | 11 | 4974 | |||
XM_006715462.3 | XP_006715525.1 | 12 | 5389 | |||
XM_011535759.2 | XP_011534061.1 | 11 | 5339 | |||
XM_011535762.2 | XP_011534064.1 | 9 | 4613 | |||
XM_017010764.1 | XP_016866253.1 | 12 | 5528 | |||
XM_017010765.1 | XP_016866254.1 | 13 | 5783 | |||
XM_017010767.1 | XP_016866256.1 | 11 | 5339 | |||
XM_017010768.1 | XP_016866257.1 | 11 | 5153 | |||
XM_017010769.1 | XP_016866258.1 | 11 | 5041 | |||
XM_017010770.1 | XP_016866259.1 | 13 | 5732 | |||
XM_017010771.1 | XP_016866260.1 | 9 | 4618 | |||
XM_017010772.1 | XP_016866261.1 | 10 | 5077 |
Gene Symbol: DHRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126383.1 | - | 1 | 1121 |
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080432.2 | NP_001073901.1 | 9 | 3685 | ||
XM_011523313.2 | XP_011521615.1 | 9 | 3679 | |||
GenBank mRNA | AB051539.1 | - | 9 | 3467 | ||
AK096554.1 | - | 9 | 3490 | |||
BC001284.1 | - | 1 | 739 | |||
BC003583.1 | - | 3 | 2194 | |||
BC030798.1 | - | 3 | 2451 |
Gene Symbol: GJC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR933719.1 | - | 3 | 1747 |
Gene Symbol: GTF2H2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537982.1 | - | 7 | 413 |
Gene Symbol: HELLPAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JX088243.1 | - | 1 | 96560 |
Gene Symbol: LOC100996574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015413.1 | XP_016870902.1 | 1 | 1171 |
Gene Symbol: NKAPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537594.1 | - | 1 | 3515 |
Gene Symbol: PDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209241.1 | - | 2 | 1075 |
Gene Symbol: PNMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007257.5 | NP_009188.1 | 3 | 2122 | ||
XM_011544365.2 | XP_011542667.1 | 3 | 2221 | |||
GenBank mRNA | AB020690.1 | - | 1 | 1529 | ||
AF083115.1 | - | 1 | 1111 | |||
AK055325.1 | - | 3 | 2064 | |||
BC036489.1 | - | 3 | 2052 | 5262276 | ||
BC047515.1 | - | 3 | 2027 | 5294654 | ||
BC062301.1 | - | 1 | 1603 | 6580976 |
Gene Symbol: RHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X63097.1 | - | 10 | 1647 |
Gene Symbol: SAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033503.2 | NP_001028675.1 | 8 | 4036 | ||
NM_016103.3 | NP_057187.1 | 7 | 3917 | |||
XM_017009521.1 | XP_016865010.1 | 7 | 3924 |
Gene Symbol: SEPHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125066.1 | - | 1 | 469 |
Gene Symbol: SERPINB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122895.1 | - | 1 | 701 | ||
AK128637.1 | - | 1 | 2127 |
Gene Symbol: SLC46A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097194.1 | - | 1 | 958 |
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127482.2 | NP_001120954.1 | 4 | 2418 | ||
NM_020456.3 | NP_065189.1 | 5 | 2535 | |||
NR_023351.2 | 3 | 2251 | ||||
GenBank mRNA | AF334405.1 | - | 5 | 2374 |
Gene Symbol: STON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033104.3 | NP_149095.2 | 6 | 10089 | ||
XM_006720279.1 | XP_006720342.1 | 6 | 7806 | |||
XM_011537242.1 | XP_011535544.1 | 7 | 7977 | |||
GenBank mRNA | BX648249.1 | - | 1 | 3767 |
Gene Symbol: TIMM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC005236.1 | - | 2 | 220 |
Gene Symbol: WHAMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080435.2 | NP_001073904.1 | 10 | 3083 | ||
XM_005272421.4 | XP_005272478.1 | 10 | 2699 | |||
XM_005272422.3 | XP_005272479.1 | 10 | 2446 | |||
XM_005272423.4 | XP_005272480.1 | 10 | 2685 | |||
XM_011521232.2 | XP_011519534.1 | 8 | 2895 | |||
GenBank mRNA | AB075851.1 | - | 10 | 3069 | ||
AK097947.1 | - | 2 | 1207 | |||
AK126887.1 | - | 10 | 3085 |
Gene Symbol: ZNF131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310735.1 | - | 7 | 610 |