siRNA Details
Gene Symbol: FDPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135821.1 | NP_001129293.1 | 3 | 406 | ||
NM_001135822.1 | NP_001129294.1 | 2 | 263 | |||
NM_001242824.1 | NP_001229753.1 | 2 | 284 | |||
NM_002004.3 | NP_001995.1 | 3 | 461 | |||
XM_005244962.1 | XP_005245019.1 | 2 | 174 | |||
XM_005244963.1 | XP_005245020.1 | 2 | 229 | |||
GenBank mRNA | AK021828.1 | - | 1 | 1197 | ||
AK022841.1 | - | 1 | 1172 | |||
AK291084.1 | - | 3 | 262 | |||
BC010004.2 | - | 2 | 250 | 4132071 | ||
BP370037.1 | - | 2 | 288 | |||
CN346026.1 | - | 2 | 233 | |||
D14697.1 | - | 3 | 356 | |||
DA610405.1 | - | 2 | 229 | |||
J05262.1 | - | 1 | 50 | |||
M29863.1 | - | 1 | 24 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
