Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021006.5 | NP_066286.1 | 1, 2 | 166 | ||
GenBank mRNA | AV739733.1 | - | 1, 2 | 210 | ||
BC027888.1 | - | 1, 2 | 154 | 5222247 | ||
BC107710.1 | - | 1, 2 | 170 | 6699133 | ||
BC146914.1 | - | 1, 2 | 117 | 9056842 |
Gene Symbol: CCL3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001437.3 | NP_001001437.2 | 1, 2 | 156 | ||
GenBank mRNA | BC007783.2 | - | 1, 2 | 151 | 4308472 | |
BC146919.1 | - | 1, 2 | 117 | 9056847 | ||
BC147008.1 | - | 1, 2 | 117 | 9056936 | ||
BC147013.1 | - | 1, 2 | 117 | 9056941 | ||
BT006783.1 | - | 1, 2 | 81 | |||
CF128666.1 | - | 1, 2 | 135 |