Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM49B SIRNA Assays ›
Gene Symbol:
FAM49B
Gene Name:
family with sequence similarity 49 member B
Gene Aliases:
BM-009, L1
Chromosome Location:
Chr.8: 129839593 - 130016697 on Build GRCh38
UniGene ID:
Hs.126941
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24824
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM49B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256763.1
NP_001243692.1
12, 13
1320
NM_016623.4
NP_057707.3
11, 12
1233
NR_046359.1
14, 15
1586
NR_046360.1
14, 15
1496
NR_046361.1
13, 14
1486
XM_005250966.3
XP_005251023.1
10, 11
1089
XM_011517108.2
XP_011515410.1
13, 14
1382
XM_011517109.2
XP_011515411.1
13, 14
1154
XM_011517110.2
XP_011515412.1
12, 13
1294
XM_011517111.2
XP_011515413.1
12, 13
1112
XM_011517112.2
XP_011515414.1
14, 15
1337
XM_011517113.2
XP_011515415.1
13, 14
1348
XM_011517114.2
XP_011515416.1
13, 14
1254
XM_011517115.2
XP_011515417.1
12, 13
1261
XM_011517116.2
XP_011515418.1
13, 14
1295
XM_011517117.2
XP_011515419.1
12, 13
1264
XM_011517118.2
XP_011515420.1
12, 13
1208
XM_011517120.2
XP_011515422.1
11, 12
1207
XM_011517121.2
XP_011515423.1
14, 15
1219
XM_011517122.2
XP_011515424.1
12, 13
1298
XM_011517123.2
XP_011515425.1
12, 13
1071
XM_011517124.2
XP_011515426.1
11, 12
1186
XM_011517125.2
XP_011515427.1
11, 12
1125
XM_017013539.1
XP_016869028.1
14, 15
1341
XM_017013540.1
XP_016869029.1
14, 15
1200
XM_017013541.1
XP_016869030.1
12, 13
1328
XM_017013542.1
XP_016869031.1
14, 15
1378
XM_017013543.1
XP_016869032.1
14, 15
1478
XM_017013544.1
XP_016869033.1
14, 15
1260
XM_017013545.1
XP_016869034.1
15, 16
1524
XM_017013546.1
XP_016869035.1
13, 14
1291
XM_017013547.1
XP_016869036.1
15, 16
1383
XM_017013548.1
XP_016869037.1
13, 14
1387
XM_017013549.1
XP_016869038.1
13, 14
1250
XM_017013550.1
XP_016869039.1
12, 13
1301
XM_017013551.1
XP_016869040.1
14, 15
1296
XM_017013552.1
XP_016869041.1
14, 15
1341
XM_017013553.1
XP_016869042.1
13, 14
1154
XM_017013554.1
XP_016869043.1
15, 16
1366
XM_017013555.1
XP_016869044.1
12, 13
1321
XM_017013556.1
XP_016869045.1
12, 13
1067
XM_017013557.1
XP_016869046.1
13, 14
1113
XM_017013558.1
XP_016869047.1
13, 14
1258
XM_017013559.1
XP_016869048.1
13, 14
1117
XM_017013560.1
XP_016869049.1
11, 12
1232
XM_017013561.1
XP_016869050.1
11, 12
984
XM_017013562.1
XP_016869051.1
12, 13
1171
GenBank mRNA
AF208851.1
-
13, 14
1174
AK001226.1
-
7, 8
739
AK130372.1
-
11, 12
1120
AY598320.1
-
9, 10
919
BC003599.1
-
12, 13
1099
3501828
BC016345.1
-
13, 14
1240
4041187
BC017297.2
-
14, 15
1468
5017493
CR749628.1
-
10, 11
3777
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001126267, XM_008765501, XM_006241634, XM_006241633, XM_006241632, and others.This siRNA also targets the following transcripts in Mouse: NM_144846, AK075613, BC011343, AK028062.
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-sjrww:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0