Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288713.1 | NP_001275642.1 | 19 | 2267 | ||
NM_001288714.1 | NP_001275643.1 | 17 | 2012 | |||
NM_006157.4 | NP_006148.2 | 18 | 2183 | |||
NM_201551.2 | NP_963845.1 | 17 | 2042 | |||
GenBank mRNA | AK093338.1 | - | 4 | 1170 | ||
AK127805.1 | - | 19 | 2247 | |||
AK299558.1 | - | 14 | 1584 | |||
AK303736.1 | - | 17 | 1981 | |||
AK313445.1 | - | 18 | 2163 | |||
BC069674.1 | - | 18 | 2074 | 7262625 | ||
BC096100.1 | - | 17 | 1932 | 40001476 | ||
BC096101.1 | - | 18 | 2073 | 40001477 | ||
BC096102.3 | - | 18 | 2073 | 40001478 | ||
BC096103.3 | - | 17 | 1932 | 40001479 | ||
D83017.1 | - | 18 | 2112 | |||
JX565026.1 | - | 17 | 1891 | |||
U57523.1 | - | 8 | 865 |