Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080495.2 | NP_001073964.2 | 11 | 4226 | ||
XM_017012728.1 | XP_016868217.1 | 11 | 6723 | |||
XM_017012729.1 | XP_016868218.1 | 10 | 4430 | |||
XM_017012730.1 | XP_016868219.1 | 10 | 4045 | |||
XM_017012731.1 | XP_016868220.1 | 10 | 4049 | |||
XM_017012732.1 | XP_016868221.1 | 10 | 4046 | |||
XM_017012733.1 | XP_016868222.1 | 9 | 3905 | |||
XM_017012734.1 | XP_016868223.1 | 10 | 4931 | |||
XM_017012735.1 | XP_016868224.1 | 10 | 3933 | |||
XM_017012736.1 | XP_016868225.1 | 10 | 4520 | |||
XM_017012737.1 | XP_016868226.1 | 10 | 6293 | |||
XM_017012738.1 | XP_016868227.1 | 10 | 3815 | |||
XM_017012739.1 | XP_016868228.1 | 10 | 4050 | |||
XM_017012740.1 | XP_016868229.1 | 10 | 4050 | |||
GenBank mRNA | AB058759.1 | - | 1 | 511 | ||
AK027341.1 | - | 2 | 686 |