Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257194.1 | NP_001244123.1 | 15 | 4459 | ||
NM_001257195.1 | NP_001244124.1 | 13 | 4035 | |||
NM_017415.2 | NP_059111.2 | 15 | 4284 | |||
GenBank mRNA | AF208068.1 | - | 15 | 3962 | ||
AF208069.1 | - | 13 | 4066 | |||
AF208070.1 | - | 15 | 4453 |