Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321170.1 | NP_001308099.1 | 13 | 979 | ||
NM_138501.5 | NP_612510.1 | 12 | 905 | |||
NR_038103.1 | 12 | 937 | ||||
NR_038104.1 | 12 | 970 | ||||
XM_006722945.1 | XP_006723008.1 | 13 | 1023 | |||
XM_011528442.2 | XP_011526744.1 | 12 | 853 | |||
GenBank mRNA | AF038958.1 | - | 12 | 851 | ||
AF038959.1 | - | 7 | 334 | |||
AF222742.1 | - | 12 | 826 | |||
AK001416.1 | - | 12 | 869 | |||
AK094087.1 | - | 13 | 946 | |||
AK299134.1 | - | 13 | 943 | |||
AK315412.1 | - | 12 | 869 | |||
BC000174.2 | - | 11 | 765 | |||
BC002425.1 | - | 12 | 837 | 3163592 | ||
BC005952.1 | - | 12 | 848 | 4249841 | ||
BC007801.2 | - | 12 | 855 | 4128702 | ||
BC013881.2 | - | 12 | 853 | 3928698 | ||
BT007179.1 | - | 11 | 775 | |||
CR456892.1 | - | 11 | 775 |