Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005577.2 | NP_005568.2 | 35 | 5609 | ||
GenBank mRNA | X06290.1 | - | 13058 |
Gene Symbol: PLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000301.3 | NP_000292.1 | 14 | 1884 | ||
GenBank mRNA | AK298338.1 | - | 10 | 1240 | ||
AK312966.1 | - | 14 | 1835 | |||
BC060513.1 | - | 14 | 1810 | 4609795 | ||
CR749293.1 | - | 2 | 2199 | |||
K02922.1 | - | 7 | 899 | |||
M74220.1 | - | 14 | 1821 | |||
X05199.1 | - | 14 | 1826 |