Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291504.1 | NP_001278433.1 | 4 | 731 | ||
NM_001291505.1 | NP_001278434.1 | 3 | 653 | |||
NM_001316325.1 | NP_001303254.1 | 5 | 876 | |||
NM_006241.7 | NP_006232.1 | 4 | 731 | |||
XM_006713682.3 | XP_006713745.1 | 4 | 678 | |||
GenBank mRNA | AJ133812.1 | - | 4 | 358 | ||
AY063767.1 | - | 4 | 357 | |||
BC007655.2 | - | 4 | 580 | 3346573 | ||
BI560631.1 | - | 5 | 638 | |||
X78873.1 | - | 4 | 358 | |||
Z29646.1 | - | 4 | 355 |
Gene Symbol: PPP1R2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038443.1 | 1 | 380 | |||
GenBank mRNA | BC066922.1 | - | 1 | 380 | ||
JF438008.1 | - | 1 | 432 |