Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018012.3 | NP_060482.2 | 7, 8 | 2089 | ||
GenBank mRNA | AK293964.1 | - | 7, 8 | 2004 | ||
AK297303.1 | - | 7, 8 | 2085 | |||
AY923834.1 | - | 7, 8 | 2090 | |||
BC035896.1 | - | 4, 5 | 610 |