Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OBSCN SIRNA Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
UniGene ID:
Hs.650039
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
251366
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OBSCN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098623.2
NP_001092093.2
33
9019
NM_001271223.2
NP_001258152.2
38
10306
NM_052843.3
NP_443075.3
33
9019
XM_005273287.4
XP_005273344.1
37
10110
XM_005273291.4
XP_005273348.1
37
10110
XM_005273298.4
XP_005273355.1
37
10110
XM_005273307.4
XP_005273364.1
37
10110
XM_006711822.3
XP_006711885.1
37
10110
XM_006711823.3
XP_006711886.1
37
10110
XM_006711827.3
XP_006711890.1
37
10109
XM_006711829.3
XP_006711892.1
36
9833
XM_011544297.2
XP_011542599.1
37
10110
XM_011544299.2
XP_011542601.1
36
9834
XM_017002443.1
XP_016857932.1
37
10110
XM_017002444.1
XP_016857933.1
37
10073
XM_017002445.1
XP_016857934.1
37
10110
XM_017002446.1
XP_016857935.1
37
10110
XM_017002447.1
XP_016857936.1
37
10110
XM_017002448.1
XP_016857937.1
37
9960
XM_017002449.1
XP_016857938.1
37
10111
XM_017002450.1
XP_016857939.1
37
10111
XM_017002451.1
XP_016857940.1
37
10111
XM_017002452.1
XP_016857941.1
37
10111
XM_017002453.1
XP_016857942.1
36
9834
XM_017002454.1
XP_016857943.1
36
9834
XM_017002455.1
XP_016857944.1
36
9834
XM_017002456.1
XP_016857945.1
36
9834
XM_017002457.1
XP_016857946.1
36
9834
XM_017002458.1
XP_016857947.1
37
10111
XM_017002459.1
XP_016857948.1
37
10111
XM_017002460.1
XP_016857949.1
37
10111
XM_017002461.1
XP_016857950.1
37
10110
XM_017002462.1
XP_016857951.1
35
9558
XM_017002463.1
XP_016857952.1
35
9537
XM_017002464.1
XP_016857953.1
37
10110
XM_017002465.1
XP_016857954.1
37
10111
XM_017002466.1
XP_016857955.1
37
10111
XM_017002467.1
XP_016857956.1
37
10111
XM_017002468.1
XP_016857957.1
37
10111
XM_017002469.1
XP_016857958.1
37
10111
XM_017002470.1
XP_016857959.1
37
10112
XM_017002471.1
XP_016857960.1
37
10109
XM_017002472.1
XP_016857961.1
37
10109
XM_017002473.1
XP_016857962.1
37
10109
GenBank mRNA
AB046776.1
-
3
772
AJ002535.1
-
34
9016
AK097489.1
-
5
1253
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-86647:80/10.239.162.121:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0