Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317939.1 | NP_001304868.1 | 5 | 2341 | ||
NM_001317940.1 | NP_001304869.1 | 5 | 2247 | |||
NM_017637.5 | NP_060107.3 | 6 | 2467 | |||
XM_011517924.1 | XP_011516226.1 | 5 | 2391 | |||
XM_011517934.2 | XP_011516236.1 | 3 | 3419 | |||
XM_017014816.1 | XP_016870305.1 | 8 | 4777 | |||
XM_017014817.1 | XP_016870306.1 | 7 | 2926 | |||
XM_017014818.1 | XP_016870307.1 | 6 | 2841 | |||
XM_017014819.1 | XP_016870308.1 | 7 | 2925 | |||
XM_017014820.1 | XP_016870309.1 | 7 | 2925 | |||
XM_017014821.1 | XP_016870310.1 | 6 | 2839 | |||
XM_017014822.1 | XP_016870311.1 | 4 | 2387 | |||
XM_017014823.1 | XP_016870312.1 | 4 | 7156 | |||
XM_017014824.1 | XP_016870313.1 | 3 | 2708 | |||
XM_017014825.1 | XP_016870314.1 | 7 | 2924 | |||
XM_017014826.1 | XP_016870315.1 | 7 | 2924 | |||
XM_017014827.1 | XP_016870316.1 | 7 | 2924 | |||
XM_017014828.1 | XP_016870317.1 | 7 | 2923 | |||
XM_017014829.1 | XP_016870318.1 | 5 | 2895 | |||
GenBank mRNA | AK000050.1 | - | 1 | 251 | ||
AK092247.1 | - | 1 | 1197 | |||
AK131398.1 | - | 6 | 2349 | |||
AK299073.1 | - | 5 | 2217 | |||
AK304745.1 | - | 3 | 2009 | |||
AY438376.1 | - | 4 | 2112 | |||
BX641030.1 | - | 1 | 1497 | |||
CR933649.1 | - | 5 | 2298 |