Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010953.1 | NP_001010953.1 | 2 | 323 | ||
GenBank mRNA | BC089058.1 | - | 2 | 323 | 7315480 | |
D82867.1 | - | 2 | 353 |