Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011906.1 | NP_001011906.1 | 5 | 1007 | ||
GenBank mRNA | BC086598.1 | - | 5 | 1007 | 7313105 |