Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193335.1 | NP_001180264.1 | 2 | 461 | ||
NM_017680.4 | NP_060150.4 | 2 | 461 | |||
GenBank mRNA | AF316824.1 | - | 1 | 179 | ||
AK027359.1 | - | 2 | 264 | |||
AK172737.1 | - | 2 | 290 | |||
AK304501.1 | - | 2 | 268 | |||
AK308107.1 | - | 2 | 374 | |||
AY029191.1 | - | 2 | 394 | |||
AY358329.1 | - | 2 | 394 | |||
BC022059.1 | - | 2 | 412 | |||
BC063114.1 | - | 2 | 426 | 30404602 | ||
BP292032.1 | - | 2 | 458 | |||
CD110559.1 | - | 408 |