Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024979.1 | NP_001020150.1 | 6 | 490 | ||
XM_008761129.1 | XP_008759351.1 | 7 | 1021 | |||
XM_008761130.1 | XP_008759352.1 | 7 | 1071 | |||
XM_008761131.1 | XP_008759353.1 | 6 | 1007 | |||
XM_008761132.1 | XP_008759354.1 | 7 | 1185 | |||
GenBank mRNA | BC093381.1 | - | 6 | 490 | 7388663 | |
FQ233538.1 | - | 3 | 212 |