Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012978.2 | NP_001012996.1 | 3 | 510 | ||
NM_001159560.1 | NP_001153032.1 | 3 | 552 | |||
GenBank mRNA | AY833564.1 | - | 3 | 458 | ||
BC042818.1 | - | 3 | 459 | 5313992 | ||
BC092450.1 | - | 3 | 430 | 30720892 | ||
BC106955.2 | - | 3 | 511 | 40035908 |