Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018088.2 | NP_001018098.1 | 76 | 10405 | ||
NM_017684.4 | NP_060154.3 | 74 | 10276 | |||
NM_018080.3 | NP_060550.2 | 74 | 10276 | |||
NM_020821.2 | NP_065872.1 | 76 | 10405 | |||
GenBank mRNA | AJ608770.1 | - | 74 | 10259 | ||
AJ608771.1 | - | 76 | 10388 | |||
AJ626860.1 | - | 74 | 10259 | |||
AJ626861.1 | - | 76 | 10388 | |||
AK000143.1 | - | 4 | 404 | |||
AK123851.1 | - | 9 | 963 | |||
AK128650.1 | - | 30 | 4819 | |||
AK304408.1 | - | 11 | 1310 | |||
AL833377.1 | - | 10 | 1006 | |||
BC069387.1 | - | 4 | 404 | |||
BX648766.1 | - | 12 | 1357 | |||
CR936716.1 | - | 25 | 3887 |