Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008829.1 | NP_001008829.1 | 5 | 987 | ||
GenBank mRNA | BC088465.1 | - | 5 | 1003 | 7127909 | |
FQ230749.1 | - | 2 | 325 | |||
FQ233418.1 | - | 5 | 1013 | |||
X90376.1 | - | 4 | 921 |