Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024952.2 | NP_001020123.1 | 5, 6 | 1214 | ||
GenBank mRNA | AK139648.1 | - | 5, 6 | 1186 | ||
AK173335.1 | - | 5, 6 | 1160 | |||
AY948287.1 | - | 5, 6 | 1155 | |||
BC138663.1 | - | 4, 5 | 938 | 8861684 |