Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317177.1 | NP_001304106.1 | 9 | 919 | ||
NM_017812.3 | NP_060282.1 | 8 | 904 | |||
NR_133671.1 | 11 | 1285 | ||||
GenBank mRNA | AK000427.1 | - | 8 | 820 | ||
AK129533.1 | - | 8 | 785 | |||
AK310236.1 | - | 11 | 1197 | |||
AL516679.3 | - | 9 | 816 | |||
BC011596.2 | - | 8 | 826 | 3346346 | ||
BC014839.1 | - | 8 | 828 | 2901125 |