Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198961.2 | NP_945199.1 | 6 | 1721 | ||
GenBank mRNA | AJ543405.1 | - | 6 | 1723 |
Gene Symbol: Vmn2r39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105071.1 | NP_001098541.1 | 6 | 1729 |
Gene Symbol: Vmn2r50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105178.1 | NP_001098648.1 | 6 | 1729 |
Set Membership: |
G-protein coupled receptor |