Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024205.2 | NP_001019376.1 | 4 | 4317 | ||
GenBank mRNA | AK003251.1 | - | 1 | 487 | ||
AK122494.1 | - | 4 | 4288 | |||
AK142954.1 | - | 4 | 4292 |