Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMKLB SIRNA Assays ›
Gene Symbol:
RIMKLB
Gene Name:
ribosomal modification protein rimK like family member B
Gene Aliases:
FAM80B, NAAGS, NAAGS-I
Chromosome Location:
Chr.12: 8681606 - 8783098 on Build GRCh38
UniGene ID:
Hs.504670
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
258574
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMKLB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_123740.1
7
1201
XM_006719118.2
XP_006719181.1
6
5701
XM_006719127.2
XP_006719190.1
7
1578
XM_011520776.1
XP_011519078.1
6
5701
XM_011520777.1
XP_011519079.1
6
5701
XM_011520778.2
XP_011519080.1
6
5701
XM_011520779.2
XP_011519081.1
6
5701
XM_011520780.1
XP_011519082.1
7
1578
XM_011520783.1
XP_011519085.1
7
1578
XM_011520790.1
XP_011519092.1
8
2265
XM_017019677.1
XP_016875166.1
6
5701
XM_017019678.1
XP_016875167.1
6
5701
XM_017019679.1
XP_016875168.1
6
5701
XM_017019680.1
XP_016875169.1
7
1578
XM_017019681.1
XP_016875170.1
7
2401
XM_017019682.1
XP_016875171.1
6
5580
XM_017019683.1
XP_016875172.1
7
6388
XM_017019684.1
XP_016875173.1
6
5324
XM_017019685.1
XP_016875174.1
7
6103
XM_017019686.1
XP_016875175.1
6
5338
XM_017019687.1
XP_016875176.1
7
5702
XM_017019688.1
XP_016875177.1
6
5842
XM_017019689.1
XP_016875178.1
6
5391
XM_017019690.1
XP_016875179.1
6
5202
XM_017019691.1
XP_016875180.1
7
1457
XM_017019692.1
XP_016875181.1
7
1435
XM_017019693.1
XP_016875182.1
7
1457
XM_017019694.1
XP_016875183.1
7
1462
XM_017019695.1
XP_016875184.1
7
1231
XM_017019696.1
XP_016875185.1
8
2265
XM_017019697.1
XP_016875186.1
7
1152
GenBank mRNA
BC033793.1
-
7
1198
5139256
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
ATP binding
metal ion binding
N-acetyl-L-aspartate-L-glutamate ligase activity
citrate-L-glutamate ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f2ldd:80/10.239.165.14:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0