Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039081.2 | NP_001034170.1 | 5 | 473 | ||
NM_001286221.1 | NP_001273150.1 | 7 | 604 | |||
NM_001286222.1 | NP_001273151.1 | 7 | 658 | |||
NM_001286223.1 | NP_001273152.1 | 5 | 454 | |||
NM_027696.3 | NP_081972.2 | 7 | 661 | |||
GenBank mRNA | AK129405.1 | - | 7 | 581 | ||
AK138876.1 | - | 5 | 474 | |||
AK143201.1 | - | 5 | 579 | |||
AK168693.1 | - | 5 | 454 | |||
AY769435.1 | - | 6 | 453 | |||
AY769436.1 | - | 5 | 376 | |||
AY769437.1 | - | 5 | 426 | |||
AY769438.1 | - | 7 | 526 | |||
BC043930.1 | - | 7 | 586 | |||
BC108273.1 | - | 5 | 440 | 30080243 | ||
BC130227.1 | - | 5 | 425 | 40141863 | ||
BQ562973.1 | - | 2 | 51 | |||
CN538186.1 | - | 2 | 74 |