Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp54a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053871.1 | NP_446323.1 | 2 | 288 | ||
XM_006240097.2 | XP_006240159.1 | 2 | 300 | |||
GenBank mRNA | BC079117.1 | - | 2 | 288 | 7126297 | |
FQ211580.1 | - | 2 | 299 | |||
FQ225417.1 | - | 2 | 302 |